Human Recombinant VISFATIN (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Pleiotrophin (PTN) is a heparin-binding growth factor that has mitogenic effects on fibroblast, epithelial, and endothelial cells. PTN is made by many tissues, but is predominantly secreted by nervous tissue during development. PTN induces neurite outgrowth and is involved in tumor growth and metastasis.
- High quality
- Low endotoxin
- Affordable
PTN binds with low affinity to the cell surface receptor nucleolin to inhibit HIV-1 infection. PNT also binds the receptor protein tyrosine phosphatase type Z (PTPRZ), syndecan-3, and anaplastic lymphoma kinase (ALK) receptors.
Delivery: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P43490
- Reconstitution instructions:Sterile 20 mM HCl at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:PBEF
- UniProtKB:P43490
- Protein/peptide name:Visfatin
- Purity:≥90%
- Molecular weight:52,7 kDa
- Sequence:MPPNTSKVYS YFECREKKTE NSKLRKVKYE ETVFYGLQYI LNKYLKGKVV TKEKIQEAKD VYKEHFQDDV FNEKGWNYIL EKYDGHLPIE IKAVPEGFVI PRGNVLFTVE NTDPECYWLT NWIETILVQS WYPITVATNS REQKKILAKY LLETSGNLDG LEYKLHDFGY RGVSSQETAG IGASAHLVNF KGTDTVAGLA LIKKYYGTKD PVPGYSVPAA EHSTITAWGK DHEKDAFEHI VTQFSSVPVS VVSDSYDIYN ACEKIWGEDL RHLIVSRSTQ APLIIRPDSG NPLDTVLKVL EILGKKFPVT ENSKGYKLLP PYLRVIQGDG VDINTLQEIV EGMKQKMWSI ENIAFGSGGG LLQKLTRDLL NCSFKCSYVV TNGLGINVFK DPVADPNKRS KKGRLSLHRT PAGNFVTLEE GKGDLEEYGQ DLLHTVFKNG KVTKSYSFDE IRKNAQLNIE LEAAHH
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT