2-(2-Aminoethyl)pyridine
ICNA0520936910
:
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
:
:122.17 g/mol
:49…50 °C (0.03 mmHg)
:49…50 °C (0,03 mmHg)
:1,02 g/cm³ (20 °C)
:Ambient
:220-295-1
:2706-56-1
:C₇H₁₀N₂
:MFCD00006367