
2,4-Dihydroxypyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:626-03-9
:84719-31-3
Formula:C₅H₅NO₂
MDL Number:MFCD00006273
:111.10 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:∼ 267 °C (dec.)
Storage Temperature:Ambient
EINECS:210-924-8
:GHS07