2-(Methylamino)pyridine, Sigma-Aldrich®
SIAL210137-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:108.14 g/mol
:60 °C (0.2 mmHg)
:15 °C
:1.0707 g/cm³ (17.7 °C)
:224-997-9
:2810
:6.1,III
:M-Clarity™ quality level = MQ100
:4597-87-9
:C₆H₈N₂
:MFCD00006250