Order Entry
Germany
ContactUsLinkComponent
4-(Dimethylamino)pyridine
4-(Dimethylamino)pyridine
Catalog # MOLE32942968-100G
Supplier:  Molekula
CAS Number:  
4-(Dimethylamino)pyridine
Catalog # MOLE32942968-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :122.17 g/mol
Boiling Pt:195 °C (1013 hPa)
Melting Pt:109…111 °C
Flash Pt:124 °C
EINECS:214-353-5
UN:2811
ADR:6.1,I
 :GHS06
CAS Number:1122-58-3
Formula:C₇H₁₀N₂
MDL Number:MFCD00006418