Order Entry
Germany
ContactUsLinkComponent
2-Fluoro-3-nitropyridine
2-Fluoro-3-nitropyridine
Catalog # FLUO33250-5G
Supplier:  FLUOROCHEM
CAS Number:  
2-Fluoro-3-nitropyridine
Catalog # FLUO33250-5G
Supplier:  FLUOROCHEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :142.09 g/mol
CAS Number:1480-87-1
Formula:C₅H₃FN₂O₂
MDL Number:MFCD03095068