Order Entry
Germany
Orders LinkContactUsLinkComponent
Nicotinamide

Nicotinamide

100 µl
Catalog # UPST19-171
Irritant
Marketsource
Supplier:   UPSTATE TECHNOLOGY
Nicotinamide
Catalog # UPST19-171
Supplier:  UPSTATE TECHNOLOGY
Marketsource
CAS Number:  98-92-0
Nicotinamide

100 µl

CAS Number:  98-92-0
Price
Inquire For Price
Log in for availability
1 * 100 µl
Product Details & Documents
Synonyms:  3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
  • CAS Number:98-92-0
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00006395
  • Molecular Weight:122.13 g/mol
  • Merck Index:12,06574
  • Boiling Pt:334 °C (1013 hPa)
  • Melting Pt:127…128 °C
  • Density:1.4463 g/cm³ (20 °C)
  • Flash Pt:182 °C
  • Storage Temperature:Ambient
  • EINECS:202-713-4
Specifications

Specifications

  • Pk:
    100 µl

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.