Nicotinamide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:98-92-0
:
Formula:C₆H₆N₂O
MDL Number:MFCD00006395
Merck Index:12,06574
Boiling Pt:334 °C (1013 hPa)
Melting Pt:127…128 °C
Density:1.4463 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:182 °C
:122.13 g/mol
EINECS:202-713-4
:GHS07
Storage Temperature:Ambient