Nicotinamide, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Catalog # 5.00299.9020
Supplier: Merck
CAS Number:
Specifications
- Pack type:Cardboard carton
- Pk:20 kg
- Seq:4241
Specifications
About this item
:
Merck Index:12,06574
Boiling Pt:334 °C (1013 hPa)
Melting Pt:127…128 °C
Density:1.4463 g/cm³ (20 °C)
Flash Pt:182 °C
:122.13 g/mol
Storage Temperature:Ambient
EINECS:202-713-4
:GHS07
CAS Number:98-92-0
Formula:C₆H₆N₂O
MDL Number:MFCD00006395