Nicotinamide ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 98-92-0 Molecular Formula: C6H6N2O Formula Weight: 122.13 Flash Point: 182°(360°F) Physical Form: Powder Density: 1.40 Appearance: White Odour: Odourless Melting Point: 128-132° Beilstein Reference: 383619 Merck Reference: 14,6523 MDL No.: MFCD00006395
The amide of nicotinic acid (vitamin B-3)
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
- CAS Number:98-92-0
- Formula:C₆H₆N₂O
- MDL Number:MFCD00006395
- Molecular Weight:122.13 g/mol
- Merck Index:14,6523
- Boiling Pt:334 °C (1013 hPa)
- Melting Pt:128…132 °C
- Density:1.40
- Flash Pt:182 °C (360 °F)
- Storage Temperature:Ambient
- EINECS:202-713-4
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 98-92-0 Molecular Formula: C6H6N2O Formula Weight: 122.13 Flash Point: 182°(360°F) Physical Form: Powder Density: 1.40 Appearance: White Odour: Odourless Melting Point: 128-132° Beilstein Reference: 383619 Merck Reference: 14,6523 MDL No.: MFCD00006395
The amide of nicotinic acid (vitamin B-3)
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
- CAS Number:98-92-0
- Formula:C₆H₆N₂O
- MDL Number:MFCD00006395
- Molecular Weight:122.13 g/mol
- Merck Index:14,6523
- Boiling Pt:334 °C (1013 hPa)
- Melting Pt:128…132 °C
- Density:1.40
- Flash Pt:182 °C (360 °F)
- Storage Temperature:Ambient
- EINECS:202-713-4



