4-Amino-2-chloropyridine
Catalog # CHMPFL19487.POR
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
:128.56 g/mol
Boiling Pt:300 °C (1013 hPa)
Melting Pt:87…89 °C
EINECS:238-403-0
:GHS07
CAS Number:14432-12-3
Formula:C₅H₅ClN₂
MDL Number:MFCD00060089