4-Amino-2-chloropyridine 97%
:14432-12-3
:
:C₅H₅ClN₂
:MFCD00060089
:128.56 g/mol
:300 °C (1013 hPa)
:92…94 °C
:Ambient
:238-403-0
:2811
:6.1,III
:GHS06
- Appearance:Light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:89 to 94 °C
- Titration with HClO4:≥96.0 %