2-Chloronicotinonitrile
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:6602-54-6
:
Formula:C₆H₃ClN₂
MDL Number:MFCD00014628
:138.56 g/mol
Boiling Pt:112 °C (1 mmHg)
Melting Pt:107…108 °C
EINECS:229-550-1
UN:3439
ADR:6.1,III