Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Pyrene 98%

Environment
Pyrene 98%
Certificates
Pyrene 98%

3 Options Available Below

Product Details & Documents
  • CAS Number:129-00-0
  • Formula:C₁₆H₁₀
  • MDL Number:MFCD00004136
  • Molecular Weight:202.26 g/mol
  • Merck Index:12,08147
  • Boiling Pt:393 °C (760 mmHg)
  • Melting Pt:148…152 °C
  • Density:1,27 g/cm³ (20 °C)
  • Flash Pt:210 °C
  • Storage Temperature:Ambient
  • EINECS:204-927-3
  • UN:3077
  • ADR:9,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Light yellow to yellow to yellow-green Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    147 to 152 °C
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 36,40 €
    36,40 €
    1 * 25 g
       36,40 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 90,80 €
    90,80 €
    1 * 100 g
       90,80 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 312,00 €
    312,00 €
    1 * 500 g
       312,00 €