Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Nitropyrene
1-Nitropyrene
1-Nitropyrene
1-Nitropyrene
Supplier:
FLUOROCHEM
Certificates
Expand image
1-Nitropyrene
Supplier:
FLUOROCHEM
About this item
CAS Number:
5522-43-0
:
Formula:
C₁₆H₉NO₂
MDL Number:
MFCD00004138
:
247.25 g/mol
Melting Pt:
156 °C
Storage Temperature:
Refrigerator
EINECS:
226-868-2