Pyrazine, (max. 1% H₂O) 99+% (dry weight)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:290-37-9
:
Formula:C₄H₄N₂
MDL Number:MFCD00006122
:80.09 g/mol
Merck Index:14,7957
Boiling Pt:113…116 °C
Melting Pt:50…56 °C
Density:1.031
Flash Pt:55 °C (131 °F)
Storage Temperature:Ambient
EINECS:206-027-6
UN:1325
ADR:4.1,III
:GHS02
:GHS07