6-Benzylaminopurine, Sigma-Aldrich®
Catalog # SIALB8023-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:225.25 g/mol
Melting Pt:230…234 °C
:M-Clarity™ quality level = MQ100
EINECS:214-927-5
CAS Number:1214-39-7
Formula:C₁₂H₁₁N₅
MDL Number:MFCD00005572