Order Entry
Germany
ContactUsLinkComponent
6-Benzylaminopurine
6-Benzylaminopurine
Catalog # MOLEM48043715
Supplier:  Molekula
CAS Number:  
6-Benzylaminopurine
Catalog # MOLEM48043715
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :225.25 g/mol
Melting Pt:230…234 °C
EINECS:214-927-5
CAS Number:1214-39-7
Formula:C₁₂H₁₁N₅
MDL Number:MFCD00005572