β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form)
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:53-84-9
:
Formula:C₂₁H₂₇N₇O₁₄P₂
MDL Number:MFCD00036253
:663.43 g/mol
Merck Index:13,06370
Melting Pt:140 to 142 °C
Storage Temperature:Refrigerator
EINECS:200-184-4