Order Entry
Germany
Orders LinkContactUsLinkComponent
Caffeine

Caffeine

Glass bottle for solids, 10 g
Catalog # UPST19-156
Irritant
Marketsource
Supplier:   UPSTATE TECHNOLOGY
Caffeine
Catalog # UPST19-156
Supplier:  UPSTATE TECHNOLOGY
Marketsource
CAS Number:  58-08-2
Caffeine
Caffeine

Glass bottle for solids, 10 g

CAS Number:  58-08-2
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS Number:58-08-2
  • Formula:C₈H₁₀N₄O₂
  • MDL Number:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck Index:13,01636
  • Melting Pt:235…239 °C
  • Density:1.23 g/cm³ (19 °C)
  • Storage Temperature:Ambient
  • EINECS:200-362-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.