Caffeine, Sigma-Aldrich®
Catalog # SIAL27602-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
Merck Index:13,01636
Melting Pt:235…239 °C
Density:1.23 g/cm³ (19 °C)
:M-Clarity™ quality level = MQ100
:194.19 g/mol
EINECS:200-362-1
:GHS07
Storage Temperature:Ambient
CAS Number:58-08-2
Formula:C₈H₁₀N₄O₂
MDL Number:MFCD00005758