Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Caffeine, Millipore®

Glass bottle for solids, 500 g
Catalog # 205548-500
Irritant
Caffeine, Millipore®
Catalog # 205548-500
CAS Number:  58-08-2
Caffeine, Millipore®
Caffeine, Millipore®

Glass bottle for solids, 500 g

CAS Number:  58-08-2
Price
149,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS Number:58-08-2
  • Formula:C₈H₁₀N₄O₂
  • MDL Number:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck Index:13,01636
  • Melting Pt:235…239 °C
  • Density:1.23 g/cm³ (19 °C)
  • Storage Temperature:Ambient
  • EINECS:200-362-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications