Order Entry
Germany
Orders LinkContactUsLinkComponent
Xanthine

Xanthine

Glass bottle for solids, 25 g
Catalog # MOLEM61668637
Irritant
Marketsource
Supplier:   Molekula
Xanthine
Catalog # MOLEM61668637
Supplier:  Molekula
Marketsource
CAS Number:  69-89-6
Xanthine

Glass bottle for solids, 25 g

CAS Number:  69-89-6
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
  • CAS Number:69-89-6
  • Formula:C₅H₄N₄O₂
  • MDL Number:MFCD00078453
  • Molecular Weight:152.11 g/mol
  • Merck Index:12,10193
  • EINECS:200-718-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.