Xanthine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 69-89-6 Molecular Formula: C5H4N4O2 Formula Weight: 152.11 Odour: Odourless Melting Point: 300° Beilstein Reference: 8733 Merck Reference: 14,10059 MDL No.: MFCD00078453
Synonyms: 1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
- CAS Number:69-89-6
- Formula:C₅H₄N₄O₂
- MDL Number:MFCD00078453
- Molecular Weight:152.11 g/mol
- Merck Index:14,10059
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:200-718-6
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 69-89-6 Molecular Formula: C5H4N4O2 Formula Weight: 152.11 Odour: Odourless Melting Point: 300° Beilstein Reference: 8733 Merck Reference: 14,10059 MDL No.: MFCD00078453
Synonyms: 1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
- CAS Number:69-89-6
- Formula:C₅H₄N₄O₂
- MDL Number:MFCD00078453
- Molecular Weight:152.11 g/mol
- Merck Index:14,10059
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:200-718-6



