Order Entry
Germany
Orders LinkContactUsLinkComponent
Xanthine 98%

Xanthine 98%

Irritant
Xanthine 98%
Certificates
Xanthine 98%

3 Options Available Below

Product Details & Documents
Synonyms:  1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
  • CAS Number:69-89-6
  • Formula:C₅H₄N₄O₂
  • MDL Number:MFCD00078453
  • Molecular Weight:152.11 g/mol
  • Merck Index:12,10193
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:200-718-6

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (105°C, 5 hrs)
  • Sulfated ash:
    ≤0.2 % (800°C)
  • HPLC:
    ≥97.5 %
  • TLC:
    Homogeneous
  • UV spectrophotometry:
    98 to 102 % (lambda max. 278 nm, pH 10)
  • Appearance of solution:
    (c=1, 1M NaOH) clear colorless to light yellow soln
  • Heavy metals (as Pb):
    ≤10 ppm
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 44,80 €
    44,80 €
    1 * 5 g
       44,80 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 149,00 €
    149,00 €
    1 * 25 g
       149,00 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 442,00 €
    442,00 €
    1 * 100 g
       442,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.