Order Entry
Germany
ContactUsLinkComponent
tert-Nonyl mercaptan (mixture of isomers) ≥97%, Sigma-Aldrich®
Flammable
Irritant
tert-Nonyl mercaptan (mixture of isomers) ≥97%, Sigma-Aldrich®
Catalog # SIAL171034-250ML
Supplier:  Merck
CAS Number:  
Flammable
Irritant
tert-Nonyl mercaptan (mixture of isomers) ≥97%, Sigma-Aldrich®
Catalog # SIAL171034-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :160.32 g/mol
Boiling Pt:188 °C (lit.)
Density:0.856 g/ml
ADR:3,III
 :M-Clarity™ quality level = MQ100
UN:3336
 :GHS02
 :GHS07
CAS Number:25360-10-5
Formula:C₉H₂₀S
MDL Number:MFCD00059963