Order Entry
Germany
ContactUsLinkComponent
Phenazine
Phenazine
Catalog # MOLE10138748-25G
Supplier:  Molekula
CAS Number:  
Phenazine
Catalog # MOLE10138748-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :180.21 g/mol
Merck Index:12,07359
Boiling Pt:339 °C (1013 hPa)
Melting Pt:172…174 °C
Density:1.26 g/cm³ (22 °C)
EINECS:202-193-9
CAS Number:92-82-0
Formula:C₁₂H₈N₂
MDL Number:MFCD00005023