Order Entry
Germany
ContactUsLinkComponent
L(+)-Ascorbic acid ≥99.9998% (metals basis), TraceSELECT™, Fluka™
L(+)-Ascorbic acid ≥99.9998% (metals basis), TraceSELECT™, Fluka™
Catalog # 05878-100G
CAS Number:  
undefined
L(+)-Ascorbic acid ≥99.9998% (metals basis), TraceSELECT™, Fluka™
Catalog # 05878-100G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Ammonium (NH₄):
    Max. 5 ppm
  • Assay:
    Min. 99.0 %
  • Specific rotation (a) 20/D; c=10 in H₂O:
    20.5 - 21.5 °
  • Silver (Ag):
    Max. 0.01 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Arsenic (As):
    Max. 0.005 ppm
  • Gold (Au):
    Max. 0.01 ppm
  • Barium (Ba):
    Max. 0.01 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.01 ppm
  • Calcium (Ca):
    Max. 0.1 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cerium (Ce):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.05 ppm
  • Cesium (Cs):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Dysprosium (Dy):
    Max. 0.01 ppm
  • Erbium (Er):
    Max. 0.01 ppm
  • Europium (Eu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Gallium (Ga):
    Max. 0.01 ppm
  • Gadolinium (Gd):
    Max. 0.01 ppm
  • Germanium (Ge):
    Max. 0.05 ppm
  • Hafnium (Hf):
    Max. 0.01 ppm
  • Mercury (Hg):
    Max. 0.01 ppm
  • Holmium (Ho):
    Max. 0.01 ppm
  • Indium (In):
    Max. 0.01 ppm
  • Iridium (Ir):
    Max. 0.01 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lanthanum (La):
    Max. 0.01 ppm
  • Lithium (Li):
    Max. 0.01 ppm
  • Lutetium (Lu):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.05 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 2 ppm
  • Niobium (nb):
    Max. 0.01 ppm
  • Neodymium (nd):
    Max. 0.01 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Palladium (Pd):
    Max. 0.01 ppm
  • Praseodymium (Pr):
    Max. 0.01 ppm
  • Platinum (Pt):
    Max. 0.01 ppm
  • Rubidium (Rb):
    Max. 0.01 ppm
  • Rhenium (Re):
    Max. 0.01 ppm
  • Rhodium (Rh):
    Max. 0.01 ppm
  • Ruthenium (Ru):
    Max. 0.01 ppm
  • Antimony (Sb):
    Max. 0.01 ppm
  • Scandium (Sc):
    Max. 0.05 ppm
  • Selenium (Se):
    Max. 0.01 ppm
  • Samarium (Sm):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.01 ppm
  • Strontium (Sr):
    Max. 0.01 ppm
  • Tantalum (Ta):
    Max. 0.01 ppm
  • Terbium (Tb):
    Max. 0.01 ppm
  • Tellurium (Te):
    Max. 0.01 ppm
  • Thorium (Th):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.05 ppm
  • Thallium (Tl):
    Max. 0.01 ppm
  • Thulium (Tm):
    Max. 0.01 ppm
  • Uran (U):
    Max. 0.01 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Tungsten (W):
    Max. 0.01 ppm
  • Yttrium (Y):
    Max. 0.01 ppm
  • Ytterbium (Yb):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Sulfate (SO₄):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :176.13 g/mol
Merck Index:13,00837
Boiling Pt:553 °C (1013 hPa)
Melting Pt:190-194 °C
Melting Pt:190…194 °C
Density:1.65 g/cm³
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328