Order Entry
Germany
Orders LinkContactUsLinkComponent
 

L(+)-Ascorbic acid, extra pure

Supplier:   Bernd Kraft
L(+)-Ascorbic acid, extra pure
Supplier:  Bernd Kraft
Certificates
L(+)-Ascorbic acid, extra pure
L(+)-Ascorbic acid, extra pure

5 Options Available Below

Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 24,70 €
    24,70 €
    1 * 100 g
       24,70 €

  • Plastic bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 76,80 €
    76,80 €
    1 * 500 g
       76,80 €

  • Plastic bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 115,00 €
    115,00 €
    1 * 1 kg
       115,00 €

  • Plastic bottle for solids
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 349,00 €
    349,00 €
    1 * 5 kg
       349,00 €

  • Plastic bottle for solids
    10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 581,00 €
    581,00 €
    1 * 10 kg
       581,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.