Order Entry
Germany
ContactUsLinkComponent
L(+)-Ascorbic acid
L(+)-Ascorbic acid
Catalog # EHERC10303000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
L(+)-Ascorbic acid
Catalog # EHERC10303000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :176.13 g/mol
Merck Index:13,00837
Boiling Pt:553 °C (1013 hPa)
Melting Pt:190…192 °C
Density:1.694 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328