Order Entry
Germany
Orders LinkContactUsLinkComponent
 

L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®

Supplier:   Avantor
L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®
Supplier:  Avantor
Certificates
L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®
L(+)-Ascorbic acid, BAKER ANALYZED® for biotechnology, J.T.Baker®

2 Options Available Below

Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications for Product Family

Specification Test Results

  • Assay (dried):
    min. 99,5 %
  • Heavy Metals (as Pb):
    max. 0,001 %
  • Iron (Fe):
    max. 5 ppm
  • Loss on Drying at 105°C:
    max. 0,1 %
  • Residue on Ignition:
    max. 0,05 %
  • Specific Rotation [alpha] 20/D (dried basis), c=10 in water:
    +20.5 - +21.5 °
  • Water Insoluble Material:
    passes test
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 71,63 €
    71,63 €
    1 * 100 g
       71,63 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 266,80 €
    266,80 €
    1 * 500 g
       266,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.