Order Entry
Germany
ContactUsLinkComponent
Cellulase
Cellulase
Catalog # SIALSAE0020-50ML
Supplier:  Merck
CAS Number:  
Cellulase
Catalog # SIALSAE0020-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Storage conditions:
    2...8 °C
  • Form:
    Aqueous solution
  • Enzyme name:
    Cellulase
  • Enzyme synonyms:
    Cellic CTec2
  • pH:
    5,0 - 5,5 (optimal pH)

Specifications

About this item

Cellulase, enzyme blend contains cellulases, ß-glucosidases, and hemicellulase, for the application of degrading cellulose to fermentable sugars.

  • Hydrolysis (units/gram): ≥1000
  • Density: 1,0 to 1,3 g/ml

This product is effective on a wide variety of pre-treated lignocellulosic bimass materials, for converting the carbohydrates in these materials into simple sugars prior to fermentation, for application in biofuels research.

Packaging: Packed in glass bottle.