Order Entry
Germany
ContactUsLinkComponent
Cobalt(II) sulphate heptahydrate, EMSURE® for analysis, Supelco®
Cobalt(II) sulphate heptahydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Cobalt(II) sulphate heptahydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    362 g/l (20 °C)
  • Melting Point:
    98 °C
  • Molar Mass:
    281.10 g/mol
  • Bulk Density:
    800 - 1100 kg/m3
  • Density:
    1.95 g/cm3 (20 °C)
  • pH:
    4 (100 g/l, H2O, 20 °C)
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

Specification Test Results

Related Information