ContactUsLinkComponent
Cobalt(II) sulphate heptahydrate 99+%, extra pure
Irritant
Health
Environment
Cobalt(II) sulphate heptahydrate 99+%, extra pure
Certificates
undefined
Irritant
Health
Environment
Cobalt(II) sulphate heptahydrate 99+%, extra pure
Certificates
Specification Test Results
  • Appearance:
    Red-brown Adhering crystalline powder or crystals
  • Titration Complexometric:
    ≥99.0 %
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm
  • Nickel (Ni):
    ≤500 ppm