Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cobalt (II) bromide
Cobalt(II) bromide, anhydrous 97%
Cobalt(II) bromide, anhydrous 97%
:
Thermo Fisher Scientific
Cobalt(II) bromide, anhydrous 97%
:
Thermo Fisher Scientific
:
7789-43-7
:
:
Br₂Co
:
MFCD00016017
:
14,2435
:
678 °C(in N{2})
:
4.909
:
218.74 g/mol
:
232-166-7
:
GHS08
:
GHS07
:
Ambient
msds_ALFA023149
Thermo Scientific Acroseal 25 Years