
Manganese(II) bromide tetrahydrate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10031-20-6
:
Formula:Br₂Mn·4H₂O
MDL Number:MFCD00149791
:286.81 g/mol
Merck Index:14,5725
Melting Pt:64.3 °C (dec.)
Density:4.38 g/cm³ (20 °C)
EINECS:236-591-9
:GHS07
Storage Temperature:Ambient