

Titanium(IV) propanolate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3087-37-4
:
Formula:C₁₂H₂₈O₄Ti
MDL Number:MFCD00009365
:284.23 g/mol
Boiling Pt:137 °C (7 hPa)
Density:1.033 g/cm³ (20 °C)
EINECS:221-411-3
UN:2413
ADR:3,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Molar Mass:284.22 g/mol
- Boiling Point:170 °C (36 hPa)
- Flash Point:38 °C
- Refractive Index:1.4962 (25 °C, 589 nm)
- Density:1.05 g/cm3 (20 °C)