Order Entry
Germany
ContactUsLinkComponent
Zirconium(IV) oxide, Sigma-Aldrich®
Irritant
Zirconium(IV) oxide, Sigma-Aldrich®
Catalog # SIAL204994-10G
Supplier:  Merck
CAS Number:  
Irritant
Zirconium(IV) oxide, Sigma-Aldrich®
Catalog # SIAL204994-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :123.22 g/mol
Merck Index:14,10180
Boiling Pt:4300 °C (1013 hPa)
Melting Pt:2680 °C
Density:5.75 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:215-227-2
 :GHS07
CAS Number:1314-23-4
Formula:ZrO₂
MDL Number:MFCD00011310