Titanium(IV) oxide, Aeroxide® P25
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13463-67-7
:1317-70-0
Formula:TiO₂
MDL Number:MFCD00011269
:79.88 g/mol
Merck Index:13,09549
Boiling Pt:2900 °C (760 mmHg)
Melting Pt:1855 °C
Density:4.23 g/cm³
Storage Temperature:Ambient
EINECS:236-675-5
Specification Test Results
- Appearance:White Powder
- Assay:≥99.5 % (on ignited substance) Typical
- Specific surface area:35 to 65 m²/g
- pH:3.5 to 4.5 (4 % dispersion)
- Acidity (HCl):≤0.3 %