Auftragserfassung
Germany
AuftragsverfolgungKontakt
Titan(III)fluorid

Titan(III)fluorid

Marketsource
Supplier:   FLUOROCHEM
Titan(III)fluorid
Supplier:  FLUOROCHEM
Marketsource
Certificates
Titan(III)fluorid

3 Options Available Below

Product Details & Documents
Synonyms:  Titantrifluorid
  • CAS Number:13470-08-1
  • Formula:F₃Ti
  • MDL Number:MFCD00042544
  • Molecular Weight:104.88 g/mol
  • Density:2,98 g/cm³ (20 °C)
  • EINECS:236-732-4
  • UN:3260
  • ADR:8,II
  •  :Density: 2.98 g/cm³ (20 °C)
Product Family Options

Product Information

  • Packung
    VE
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 561,00 €
    561,00 €
    1 * 5 g
       561,00 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.