Order Entry
Germany
Orders LinkContactUsLinkComponent
Titanium(III) fluoride

Titanium(III) fluoride

Corrosive
Irritant
Titanium(III) fluoride
Certificates
Titanium(III) fluoride
Titanium(III) fluoride

2 Options Available Below

Product Details & Documents

CAS No.: 13470-08-1 Molecular Formula: TiF3 Formula Weight: 104.90 Physical Form: Powder Density: 3.400 Sensitivity: Air and Moisture Sensitive UN No.: 3260 MDL No.: MFCD00042544

Synonyms:  Titanium trifluoride
  • CAS Number:13470-08-1
  • Formula:F₃Ti
  • MDL Number:MFCD00042544
  • Molecular Weight:104.88 g/mol
  • Density:3.400
  • Storage Temperature:Ambient
  • EINECS:236-732-4
  • UN:3260
  • ADR:8,II
  •  :Density: 2.98 g/cm³ (20 °C)
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.