

Zinc bromide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7699-45-8
:
Formula:Br₂Zn
MDL Number:MFCD00011294
Merck Index:13,10182
Boiling Pt:670…680 °C (1013 hPa)
Melting Pt:394 °C
Density:4.22 g/cm³ (20 °C)
EINECS:231-718-4
UN:3260
ADR:8,III
:M-Clarity™ quality level = MQ200
:225.2 g/mol
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:4470 g/l (20 °C)
- Melting Point:394 °C
- Molar Mass:225.2 g/mol
- Boiling Point:697 °C (1013 mbar)
- Density:4.2 g/cm3 (20 °C)
- pH:4 (H2O, 20 °C) (saturated solution)
- Hill Formula:Br2Zn
- Chemical Formula:ZnBr2
Specification Test Results
Frequently Bought Together