


Mercury(II) bromide ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:7789-47-1
:
:Br₂Hg
:MFCD00011040
:360.4 g/mol
:13,05900
:320 °C (1013 hPa)
:236 °C
:6.1 g/cm³ (20 °C)
:Ambient
:1634
:6.1,II
:M-Clarity™ quality level = MQ300
:232-169-3
:GHS06
:GHS08
:GHS09
- Assay (complexometric):≥ 99.0 %
- Insoluble matter (in methanol):≤ 0.05 %
- Chloride (Cl):≤ 0.25 %
- Residue after reduction:≤ 0.02 %
- Correspons to ACS,Reag. Ph Eur:322 °C (1013 hPa) (sublimed)
- Vapor Pressure:3.0 hPa (150 °C)
- Density:6.1 g/cm3 (25 °C)
- pH:4.5 (5 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials