


Mercury(II) bromide ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-47-1
:
Formula:Br₂Hg
MDL Number:MFCD00011040
:360.4 g/mol
Merck Index:14,5875
Boiling Pt:322…325 °C (subl.)
Melting Pt:236 °C
Density:6.05
Storage Temperature:Ambient
EINECS:232-169-3
UN:1634
ADR:6.1,II
:GHS06
:GHS08
:GHS09