

Silver(II) oxide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1301-96-8
:
Formula:AgO
MDL Number:MFCD00044285
:123.87 g/mol
Merck Index:14,8522
Melting Pt:>100 °C (dec.)
Density:7.44
Storage Temperature:Ambient
EINECS:215-098-2
UN:3085
ADR:5.1,II
:GHS03
:GHS05