Order Entry
Germany
ContactUsLinkComponent
 
Copper(II) oxide ≥99.0% (RT), powder, Puriss. p.a., Fluka™
 
Copper(II) oxide ≥99.0% (RT), powder, Puriss. p.a., Fluka™
 

 

  • Assay:
    99.0 - 101.0 %
  • Insoluble in HCl:
    Max. 200 ppm
  • Calcium (Ca):
    Max. 100 ppm
  • Cadmium (Cd):
    Max. 10 ppm
  • Cobalt (Co):
    Max. 10 ppm
  • Chromium (Cr):
    Max. 50 ppm
  • Iron (Fe):
    Max. 500 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 100 ppm
  • Manganese (Mn):
    Max. 10 ppm
  • Sodium (Na):
    Max. 500 ppm
  • Nickel (Ni):
    Max. 50 ppm
  • Lead (Pb):
    Max. 500 ppm
  • Zinc (Zn):
    Max. 100 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Total S (as SO₄):
    Max. 500 ppm
  • Total N:
    Max. 20 ppm