Copper(II) oxide ≥94%, powder, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1317-38-0
:
Formula:CuO
MDL Number:MFCD00010979
:79.55 g/mol
Merck Index:13,02674
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:1326 °C
Density:4.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-269-1
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Assay:Min. 94 %
Specification Test Results
Related Information
Certificates
Search by Lot Number750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC23144.263 spec_VWRC23144.263 spec_VWRC23144.410