Order Entry
Germany
Orders LinkContactUsLinkComponent
Copper(II) oxide, Technipur®

Copper(II) oxide, Technipur®

Plastic bottle, 1 kg
Catalog # 1.02769.1000
Environment
Supplier:   Merck
Copper(II) oxide, Technipur®
Catalog # 1.02769.1000
Supplier:  Merck
CAS Number:  1317-38-0
Copper(II) oxide, Technipur®

Plastic bottle, 1 kg

CAS Number:  1317-38-0
Price
1 290,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Number:1317-38-0
  • Formula:CuO
  • MDL Number:MFCD00010979
  • Molecular Weight:79.55 g/mol
  • Merck Index:13,02674
  • Boiling Pt:2000 °C (1013 hPa)
  • Melting Pt:1326 °C
  • Density:4.9 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-269-1
  • UN:3077
  • ADR:9,III

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.