Copper(II) oxide, granular for elementary analysis pH 7 (50 g/l, H₂O, 20 °C) (slurry), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1317-38-0
:
Formula:CuO
MDL Number:MFCD00010979
:79.55 g/mol
Merck Index:13,02674
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:1326 °C (dec.)
Density:500 kg/m3
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:215-269-1
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay (complexometric):≥ 99
- Chloride (Cl):≤ 0.005 %
- Total sulfur (as SO₄):≤ 0.015 %
- Total nitrogen (N):≤ 0.002 %
- C (Carbon):≤ 0.01 %
- Bulk density:240 - 270 g/100 ml
- Particle size (0.71-2 mm):≥ 90 %