Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Copper(II) oxide

50 kg
Catalog # HONE12845.9050
Irritant
Environment
Supplier:   Honeywell Chemicals
Copper(II) oxide
Catalog # HONE12845.9050
CAS Number:  1317-38-0
Copper(II) oxide

50 kg

CAS Number:  1317-38-0
Price
Inquire For Price
Log in for availability
1 * 50 kg
Product Details & Documents
Synonyms:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Number:1317-38-0
  • Formula:CuO
  • MDL Number:MFCD00010979
  • Molecular Weight:79.55 g/mol
  • Merck Index:13,02674
  • Boiling Pt:2000 °C (1013 hPa)
  • Melting Pt:1326 °C
  • Density:4.9 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-269-1
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    50 kg

Specifications